missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-tert-Butylbenzyl alcohol, 98%, Thermo Scientific Chemicals
$80.84 - $750.38
Chemical Identifiers
CAS | 877-65-6 |
---|---|
Molecular Formula | C11H16O |
Molecular Weight (g/mol) | 164.248 |
MDL Number | MFCD00004662 |
InChI Key | FVEINXLJOJPHLH-UHFFFAOYSA-N |
Synonym | 4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol |
PubChem CID | 13416 |
IUPAC Name | (4-tert-butylphenyl)methanol |
SMILES | CC(C)(C)C1=CC=C(C=C1)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2126006
|
Thermo Scientific Chemicals
B2126006 |
5 g |
Each for $80.84
|
|
|||||
AAB2126014
|
Thermo Scientific Chemicals
B2126014 |
25 g |
Each for $255.81
|
|
|||||
AAB2126022
|
Thermo Scientific Chemicals
B2126022 |
100 g |
Each for $750.38
|
|
|||||
Description
4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in alcohol and insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
Chemical Identifiers
877-65-6 | |
164.248 | |
FVEINXLJOJPHLH-UHFFFAOYSA-N | |
13416 | |
CC(C)(C)C1=CC=C(C=C1)CO |
C11H16O | |
MFCD00004662 | |
4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol | |
(4-tert-butylphenyl)methanol |
Specifications
877-65-6 | |
139°C to 140°C (20 mmHg) | |
C11H16O | |
MFCD00004662 | |
1860665 | |
Soluble in alcohol and insoluble in water. | |
CC(C)(C)C1=CC=C(C=C1)CO | |
164.248 | |
164.25 | |
4-tert-Butylbenzyl alcohol |
0.928 | |
128°C (262°F) | |
1.517 | |
5 g | |
4-tert-butylbenzyl alcohol, 4-tert-butyl phenyl methanol, 4-tert-butylphenyl methanol, 4-t-butylbenzyl alcohol, p-tert-butylbenzyl alcohol, 4-t-butylbenzylalcohol, benzenemethanol, 4-1,1-dimethylethyl, 4-1,1-dimethylethyl benzenemethanol, 4-tert-butylbenzylalcohol, p-t-butylbenzyl alcohol | |
FVEINXLJOJPHLH-UHFFFAOYSA-N | |
(4-tert-butylphenyl)methanol | |
13416 | |
98% |
Safety and Handling
EINECSNumber : 212-894-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only