missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylbenzyl chloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1185606

 View more versions of this product

Catalog No. AAA1185606


Explore available promotions

Description

Description

4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
Specifications

Specifications

4-tert-Butylbenzyl chloride
0.945
94° (202°F)
1.522
5 g
471673
Slightly soluble in water.
CC(C)(C)C1=CC=C(CCl)C=C1
182.69
182.7
19692-45-6
101°C to 104°C (7 mmHg)
C11H15Cl
MFCD00000918
UN1760
4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
1-tert-butyl-4-(chloromethyl)benzene
88198
99%

Chemical Identifiers

19692-45-6
182.69
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
88198
CC(C)(C)C1=CC=C(CCl)C=C1
C11H15Cl
MFCD00000918
4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
1-tert-butyl-4-(chloromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 243-228-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only