missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcatechol, 99%

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol

$80.05 - $2693.83

Identifiants chimiques

CAS 98-29-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002201
InChI Key XESZUVZBAMCAEJ-UHFFFAOYSA-N
Synonym 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
PubChem CID 7381
IUPAC Name 4-tert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC=C(O)C(O)=C1
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC154351000
FDS Afficher les documents
Thermo Scientific Chemicals
154351000
100 g Plastic bottle
chaque for $80.05
Il en reste null
 
AC154355000
FDS Afficher les documents
Thermo Scientific Chemicals
154355000
500 g Plastic bottle
chaque for $244.20
Il en reste null
 
AC154350025
FDS Afficher les documents
Thermo Scientific Chemicals
154350025
2.5 kg Plastic Bottle
chaque for $822.52
Il en reste null
 
AC154350100
FDS Afficher les documents
Thermo Scientific Chemicals
154350100
10 kg Plastic drum
chaque for $2,693.83
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

98-29-3
166.22
XESZUVZBAMCAEJ-UHFFFAOYSA-N
7381
CC(C)(C)C1=CC=C(O)C(O)=C1
C10H14O2
MFCD00002201
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
4-tert-butylbenzene-1,2-diol

Spécifications

98-29-3
100.0
1.0490g/mL
129°C
Plastic bottle
(CH3)3CC6H3(OH)2
100 g
15,54
4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
XESZUVZBAMCAEJ-UHFFFAOYSA-N
4-tert-butylbenzene-1,2-diol
7381
166.22
Technical
98.5
53.0°C to 56.0°C
285.0°C
0.7% max. Catechol (GC)
C10H14O2
MFCD00002201
06,IV,6014
1.049
Solubility in water: 0.2% (25°C). Other solubilities: soluble in hot water,very sol. in alcohol,ethers,esters and ketones,partly sol. in chlorinated and aromatic hydrocarb.,slightly sol. in aliphatics. 240g/100g acetone,80g/100g benzene;60g/100g
CC(C)(C)C1=CC=C(O)C(O)=C1
166.22
90 mm2/s (25°C)
99%
4-tert-Butylcatechol, 99%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting eff

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-653-9

missing translation for 'rtecsNumber' : UX1400000

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only