missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107900050

 View more versions of this product

Catalog No. AC107900050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-tert-Butylcyclohexanol
98-52-2
100.0
White
200°C
98.5% min. (GC)
C10H20O
(CH3)3CC6H10OH
5 g
4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
CCOQPGVQAWPUPE-UHFFFAOYSA-N
4-tert-butylcyclohexan-1-ol
7391
156.27
Powder or Granules
99%
98.5
60.0°C to 69.0°C
224.0°C
Authentic
Glass Bottle
1.447
MFCD00001473,MFCD00064952,MFCD00070476
06, I, 18
Solubility in water: <1g/L water (20 c)
CC(C)(C)C1CCC(O)CC1
156.27
8.68 mPa.s (80°C)
99%

Chemical Identifiers

98-52-2
156.27
CCOQPGVQAWPUPE-UHFFFAOYSA-N
7391
CC(C)(C)C1CCC(O)CC1
C10H20O
MFCD00001473,MFCD00064952,MFCD00070476
4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
4-tert-butylcyclohexan-1-ol
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 202-676-4

RTECSNumber : GV8750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only