missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylphenol, 97%, Thermo Scientific Chemicals

$45.20 - $116.65

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC108000050
SDS View Documents
Thermo Scientific Chemicals
108000050
5 g
Each for $45.20
 
AC108000010
SDS View Documents
Thermo Scientific Chemicals
108000010
1 kg
Each for $116.65
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-54-4
150.22
QHPQWRBYOIRBIT-UHFFFAOYSA-N
7393
4-tert-butylphenol
C10H14O
MFCD00002367
4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
CHEBI:34444
CC(C)(C)C1=CC=C(C=C1)O

Specifications

98-54-4
100.0
White to Beige
236.0°C to 238.0°C
Authentic
Glass bottle
1.4787
MFCD00002367
06, 524
15, 1588
Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in aq. alcoholic alkali,soluble in alcohol and ether
CC(C)(C)C1=CC=C(C=C1)O
150.22
2.4 mPa.s (110°C)
150.22
Flakes or Pastilles
96.0
96.0°C to 100.0°C
0.9000g/mL
113°C
96% min. (GC)
C10H14O
(CH3)3CC6H4OH
5 g
0.9
4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
QHPQWRBYOIRBIT-UHFFFAOYSA-N
4-tert-butylphenol
7393
CHEBI:34444
97%
4-tert-Butylphenol, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging fertility.

GHS P Statement
Obtain special instructions before use.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several

GHS Signal Word: Danger

EINECSNumber : 202-679-0

RTECSNumber : SJ8925000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only