Learn More
4-tert-Butylphenol, 99%
CAS: 98-54-4 | C10H14O | 150.221 g/mol
$78.73 - $305.40
Chemical Identifiers
| CAS | 98-54-4 |
|---|---|
| Molecular Formula | C10H14O |
| MDL Number | MFCD00002367 |
| InChI Key | QHPQWRBYOIRBIT-UHFFFAOYSA-N |
| Synonym | 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol |
| PubChem CID | 7393 |
| ChEBI | CHEBI:34444 |
| IUPAC Name | 4-tert-butylphenol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1587136
|
Thermo Scientific Chemicals
A1587136 |
500 g |
Each for $78.73
|
|
|||||
|
AAA158710E
|
Thermo Scientific Chemicals
A158710E |
2500 g |
Each for $305.40
|
|
|||||
Description
4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.
Solubility
Soluble in water. [8.7 g/L (20°C)]
Notes
Store at room temperature. Keep away from oxidizing agents and acids.
Chemical Identifiers
| 98-54-4 | |
| MFCD00002367 | |
| 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol | |
| CHEBI:34444 |
| C10H14O | |
| QHPQWRBYOIRBIT-UHFFFAOYSA-N | |
| 7393 | |
| 4-tert-butylphenol |
Specifications
| 98-54-4 | |
| 0.908 | |
| 113°C (235°F) | |
| C10H14O | |
| 500 g | |
| 1817334 | |
| 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol | |
| QHPQWRBYOIRBIT-UHFFFAOYSA-N | |
| 4-tert-butylphenol | |
| 7393 | |
| 150.22 | |
| 4-tert-Butylphenol |
| 97°C to 101°C | |
| 236°C to 238°C | |
| 99% | |
| MFCD00002367 | |
| UN3077 | |
| 14,1585 | |
| Soluble in water. [8.7g/L (20°C)] | |
| CC(C)(C)C1=CC=C(C=C1)O | |
| 150.221 | |
| CHEBI:34444 | |
| 99% |
Safety and Handling
GHS H Statement
H318-H317-H335-H315
P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
H315-H318-H361f
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-679-0
RTECSNumber : SJ8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only