missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenol 98.0+%, TCI America™

$31.68 - $50.33

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B023025G
View Documents
TCI America
B023025G
25 g
Each for $31.68
Only null left
 
B0230500G
View Documents
TCI America
B0230500G
500 g
Each for $50.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

98-54-4
150.221
QHPQWRBYOIRBIT-UHFFFAOYSA-N
7393
4-tert-butylphenol
C10H14O
MFCD00002367
4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
CHEBI:34444
CC(C)(C)C1=CC=C(C=C1)O

Specifications

98-54-4
White-Yellow
C10H14O
25 g
4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
CC(C)(C)C1=CC=C(C=C1)O
150.221
CHEBI:34444
≥98.0% (GC)
4-tert-Butylphenol
99°C
238°C
MFCD00002367
2430
QHPQWRBYOIRBIT-UHFFFAOYSA-N
4-tert-butylphenol
7393
150.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0503

RTECSNumber : SJ8925000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'