missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

$93.92 - $942.07

Chemical Identifiers

CAS 1798-04-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00021758
InChI Key FBIGAJNVRFKBJL-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy
PubChem CID 15718
IUPAC Name 2-(4-tert-butylphenoxy)acetic acid
SMILES CC(C)(C)C1=CC=C(C=C1)OCC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1701706
View Documents
Thermo Scientific Chemicals
A1701706
5 g N/A
 
AAA1701714
View Documents
Thermo Scientific Chemicals
A1701714
25 g N/A
 
AAA1701722
View Documents
Thermo Scientific Chemicals
A1701722
100 g N/A
 
Description

Description

Applied as a reagent for the TAC protective group in nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied as a reagent for the TAC protective group in nucleosides.

Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications

Chemical Identifiers

1798-04-5
208.257
FBIGAJNVRFKBJL-UHFFFAOYSA-N
15718
CC(C)(C)C1=CC=C(C=C1)OCC(=O)O
C12H16O3
MFCD00021758
4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy
2-(4-tert-butylphenoxy)acetic acid

Specifications

1798-04-5
MFCD00021758
4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy
FBIGAJNVRFKBJL-UHFFFAOYSA-N
2-(4-tert-butylphenoxy)acetic acid
15718
98%
C12H16O3
5 g
Sparingly Soluble (0.56g/L) (25°C),Calc.
CC(C)(C)C1=CC=C(C=C1)OCC(=O)O
208.257
208.26
4-tert-Butylphenoxyacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only