missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

$52.59 - $118.33

Chemical Identifiers

CAS 1746-23-2
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00065126
InChI Key QEDJMOONZLUIMC-UHFFFAOYSA-N
Synonym 4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
PubChem CID 15627
IUPAC Name 1-tert-butyl-4-ethenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL12356AE
View Documents
Thermo Scientific Chemicals
L12356AE
100 mL
Each for $52.59
 
AAL12356AP
View Documents
Thermo Scientific Chemicals
L12356AP
500 mL
Each for $118.33
 
Description

Description

4-tert-Butylstyrene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylstyrene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in the dark. Protect from heat. Incompatible with heat, light, free radical initiators, UV radiation. Do not store together.
Specifications

Chemical Identifiers

1746-23-2
160.26
QEDJMOONZLUIMC-UHFFFAOYSA-N
15627
CC(C)(C)C1=CC=C(C=C1)C=C
C12H16
MFCD00065126
4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
1-tert-butyl-4-ethenylbenzene

Specifications

1746-23-2
0.884
80°C (176°F)
1.525
100 mL
4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
QEDJMOONZLUIMC-UHFFFAOYSA-N
1-tert-butyl-4-ethenylbenzene
15627
94%
-38°C
219°C
C12H16
MFCD00065126
1903714
Not miscible in water.
CC(C)(C)C1=CC=C(C=C1)C=C
160.26
160.26
4-tert-Butylstyrene, stabilized with 50ppm 4-tert-butylcatechol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H332

EINECSNumber : 217-126-9

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only