missing translation for 'onlineSavingsMsg'
Learn More

(4-Thien-2-yltetrahydropyran-4-yl)methylamine, 97%, Thermo Scientific™

$168.22 - $168.22

Chemical Identifiers

CAS 906422-74-0
Molecular Formula C10H15NOS
Molecular Weight (g/mol) 197.296
MDL Number MFCD09817518
InChI Key LBEBHUWSRFXHQR-UHFFFAOYSA-N
Synonym 4-thien-2-yltetrahydropyran-4-yl methylamine, 4-thiophen-2-yl oxan-4-yl methanamine, 1-4-thiophen-2-yl oxan-4-yl methanamine, 4-thien-2-yl tetrahydro-2h-pyran-4-yl methylamine, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methanamine, 4-thiophen-2-yltetrahydropyran-4-yl methylamine, 4-2-thienyl tetrahydro-2h-pyran-4-yl methyl amine, 4-aminomethyl-4-thien-2-yl tetrahydro-2h-pyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine
PubChem CID 24229669
IUPAC Name (4-thiophen-2-yloxan-4-yl)methanamine
SMILES C1COCCC1(CN)C2=CC=CS2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
CC62213CB
SDS View Documents
Thermo Scientific Chemicals
CC62213CB
250 mg Amber Glass Bottle
Each for $168.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

906422-74-0
197.296
LBEBHUWSRFXHQR-UHFFFAOYSA-N
24229669
C1COCCC1(CN)C2=CC=CS2
C10H15NOS
MFCD09817518
4-thien-2-yltetrahydropyran-4-yl methylamine, 4-thiophen-2-yl oxan-4-yl methanamine, 1-4-thiophen-2-yl oxan-4-yl methanamine, 4-thien-2-yl tetrahydro-2h-pyran-4-yl methylamine, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methanamine, 4-thiophen-2-yltetrahydropyran-4-yl methylamine, 4-2-thienyl tetrahydro-2h-pyran-4-yl methyl amine, 4-aminomethyl-4-thien-2-yl tetrahydro-2h-pyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine
(4-thiophen-2-yloxan-4-yl)methanamine

Specifications

906422-74-0
100.0
Amber Glass Bottle
MFCD09817518
4-thien-2-yltetrahydropyran-4-yl methylamine, 4-thiophen-2-yl oxan-4-yl methanamine, 1-4-thiophen-2-yl oxan-4-yl methanamine, 4-thien-2-yl tetrahydro-2h-pyran-4-yl methylamine, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methanamine, 4-thiophen-2-yltetrahydropyran-4-yl methylamine, 4-2-thienyl tetrahydro-2h-pyran-4-yl methyl amine, 4-aminomethyl-4-thien-2-yl tetrahydro-2h-pyran, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine
C1COCCC1(CN)C2=CC=CS2
197.296
197.3
(4-Thien-2-yltetrahydropyran-4-yl)methylamine
96.5
97%
C10H15NOS
250 mg
LBEBHUWSRFXHQR-UHFFFAOYSA-N
(4-thiophen-2-yloxan-4-yl)methanamine
24229669
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents