missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(trans-4-Ethylcyclohexyl)benzonitrile 98.0+%, TCI America™

Supplier:  TCI America E07035G

Catalog No. E07035G


Explore available promotions

Description

Description

Specifications

Specifications

4-(trans-4-Ethylcyclohexyl)benzonitrile
41°C
C15H19N
3439
BBHJTCADCKZYSO-UHFFFAOYSA-N
4-(4-ethylcyclohexyl)benzonitrile
175307
≥98.0% (GC)
72928-54-2
White-Yellow
5 g
4-4-ethylcyclohexyl benzonitrile, 4-trans-4-ethylcyclohexyl benzonitrile, trans-4-4-ethylcyclohexyl benzonitrile, 4-trans-4-ethylcyclohexyl-benzonitrile, 1-cyano-4-trans-4-ethylcyclohexyl benzene, benzonitrile, 4-trans-4-ethylcyclohexyl, 4-trans-ethylcyclohexylcyanobenzene, benzonitrile, 4-4-ethylcyclohexyl, trans-4-ethylcyclohexyl-4-benzonitrile, 4-1s,4r-4-ethylcyclohexyl benzonitrile
CCC1CCC(CC1)C2=CC=C(C=C2)C#N
213.324
213.32
Crystalline Powder

Chemical Identifiers

72928-54-2
213.324
4-4-ethylcyclohexyl benzonitrile, 4-trans-4-ethylcyclohexyl benzonitrile, trans-4-4-ethylcyclohexyl benzonitrile, 4-trans-4-ethylcyclohexyl-benzonitrile, 1-cyano-4-trans-4-ethylcyclohexyl benzene, benzonitrile, 4-trans-4-ethylcyclohexyl, 4-trans-ethylcyclohexylcyanobenzene, benzonitrile, 4-4-ethylcyclohexyl, trans-4-ethylcyclohexyl-4-benzonitrile, 4-1s,4r-4-ethylcyclohexyl benzonitrile
4-(4-ethylcyclohexyl)benzonitrile
C15H19N
BBHJTCADCKZYSO-UHFFFAOYSA-N
175307
CCC1CCC(CC1)C2=CC=C(C=C2)C#N
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions