Learn More
4-(Trifluoromethoxy)anisole, 98+%, Thermo Scientific Chemicals
$89.25 - $323.33
Chemical Identifiers
CAS | 710-18-9 |
---|---|
Molecular Formula | C8H7F3O2 |
Molecular Weight (g/mol) | 192.137 |
MDL Number | MFCD00216942 |
InChI Key | NOAFZIOGGDPYKK-UHFFFAOYSA-N |
Synonym | 4-trifluoromethoxy anisole, 1-methoxy-4-trifluoromethoxy benzene, p-trifluoromethoxy anisole, 4-trifluoromethoxyanisole, benzene, 1-methoxy-4-trifluoromethoxy, 4-methoxy trifluoromethoxy benzene, hydroquinone methyl trifluoromethyl ether, trifluoro 4-methoxyphenoxy methane, pubchem3570, acmc-1bcz3 |
PubChem CID | 2724973 |
IUPAC Name | 1-methoxy-4-(trifluoromethoxy)benzene |
SMILES | COC1=CC=C(C=C1)OC(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1974606
|
Thermo Scientific Chemicals
A1974606 |
5 g |
N/A
|
|
|||||
AAA1974614
|
Thermo Scientific Chemicals
A1974614 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
710-18-9 | |
192.137 | |
NOAFZIOGGDPYKK-UHFFFAOYSA-N | |
2724973 | |
COC1=CC=C(C=C1)OC(F)(F)F |
C8H7F3O2 | |
MFCD00216942 | |
4-trifluoromethoxy anisole, 1-methoxy-4-trifluoromethoxy benzene, p-trifluoromethoxy anisole, 4-trifluoromethoxyanisole, benzene, 1-methoxy-4-trifluoromethoxy, 4-methoxy trifluoromethoxy benzene, hydroquinone methyl trifluoromethyl ether, trifluoro 4-methoxyphenoxy methane, pubchem3570, acmc-1bcz3 | |
1-methoxy-4-(trifluoromethoxy)benzene |
Specifications
710-18-9 | |
163°C to 165°C | |
C8H7F3O2 | |
MFCD00216942 | |
UN1993 | |
4-trifluoromethoxy anisole, 1-methoxy-4-trifluoromethoxy benzene, p-trifluoromethoxy anisole, 4-trifluoromethoxyanisole, benzene, 1-methoxy-4-trifluoromethoxy, 4-methoxy trifluoromethoxy benzene, hydroquinone methyl trifluoromethyl ether, trifluoro 4-methoxyphenoxy methane, pubchem3570, acmc-1bcz3 | |
COC1=CC=C(C=C1)OC(F)(F)F | |
192.137 | |
192.14 | |
4-(Trifluoromethoxy)anisole |
1.272 | |
58°C (136°F) | |
1.432 | |
5 g | |
2556809 | |
NOAFZIOGGDPYKK-UHFFFAOYSA-N | |
1-methoxy-4-(trifluoromethoxy)benzene | |
2724973 | |
≥98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only