missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethoxy)phenylacetic acid, 98%, Thermo Scientific Chemicals

$111.95 - $374.37

Chemical Identifiers

CAS 4315-07-5
MDL Number MFCD00061239
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1951203
View Documents
Thermo Scientific Chemicals
A1951203
1 g
Each for $111.95
 
AAA1951206
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1951206
5 g
Each for $374.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4315-07-5
MFCD00061239

Specifications

4315-07-5
C34H22N8Na2O10S2
1 g
diamine green b, c.i. direct green 6, disodium salt, pontamine green, dianil green, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-6-2-4'-2-4-hydroxyphenyl diazenyl 1,1'-biphenyl-4-yl diazenyl-3-2-4-nitrophenyl diazenyl-, sodium salt 1:2
C1=CC(=O)C=CC1=NNC2=CC=C(C=C2)C3=CC=C(C=C3)NN=C4C(=CC5=CC(=C(C(=C5C4=O)N)N=NC6=CC=C(C=C6)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
812.70
220.15
4-(Trifluoromethoxy)phenylacetic acid
86°C to 88°C
MFCD00061239
2695336
LQDBIEHCXRQVQI-LYAINCPBSA-L
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-[[4-[4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]phenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
44135513
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only