missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)toluene 98.0+%, TCI America™
$134.21 - $434.32
Chemical Identifiers
| CAS | 706-27-4 |
|---|---|
| Molecular Formula | C8H7F3O |
| Molecular Weight (g/mol) | 176.138 |
| MDL Number | MFCD00042413 |
| InChI Key | JUXFXYQUXNXVAA-UHFFFAOYSA-N |
| Synonym | 1-methyl-4-trifluoromethoxy benzene, 4-trifluoromethoxytoluene, 4-trifluoromethoxy toluene, 4-toluene trifluoromethyl ether, 4-methyltrifluoromethoxybenzene, benzene, 1-methyl-4-trifluoromethoxy, 4-methyl-alpha,alpha,alpha-trifluoroanisole, pubchem13486, acmc-209ofc |
| PubChem CID | 2760631 |
| IUPAC Name | 1-methyl-4-(trifluoromethoxy)benzene |
| SMILES | CC1=CC=C(C=C1)OC(F)(F)F |
Chemical Identifiers
| 706-27-4 | |
| 176.138 | |
| JUXFXYQUXNXVAA-UHFFFAOYSA-N | |
| 2760631 | |
| CC1=CC=C(C=C1)OC(F)(F)F |
| C8H7F3O | |
| MFCD00042413 | |
| 1-methyl-4-trifluoromethoxy benzene, 4-trifluoromethoxytoluene, 4-trifluoromethoxy toluene, 4-toluene trifluoromethyl ether, 4-methyltrifluoromethoxybenzene, benzene, 1-methyl-4-trifluoromethoxy, 4-methyl-alpha,alpha,alpha-trifluoroanisole, pubchem13486, acmc-209ofc | |
| 1-methyl-4-(trifluoromethoxy)benzene |
Specifications
| 706-27-4 | |
| 135°C | |
| MFCD00042413 | |
| 1993 | |
| JUXFXYQUXNXVAA-UHFFFAOYSA-N | |
| 1-methyl-4-(trifluoromethoxy)benzene | |
| 2760631 | |
| ≥98.0% (GC) | |
| 4-(Trifluoromethoxy)toluene |
| Yellow | |
| C8H7F3O | |
| 1 g | |
| 1-methyl-4-trifluoromethoxy benzene, 4-trifluoromethoxytoluene, 4-trifluoromethoxy toluene, 4-toluene trifluoromethyl ether, 4-methyltrifluoromethoxybenzene, benzene, 1-methyl-4-trifluoromethoxy, 4-methyl-alpha,alpha,alpha-trifluoroanisole, pubchem13486, acmc-209ofc | |
| CC1=CC=C(C=C1)OC(F)(F)F | |
| 176.138 | |
| 176.14 | |
| Liquid |
Safety and Handling
TSCA : No