missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-(Trifluoromethyl)acetanilide, 98+%, Thermo Scientific Chemicals
$145.36 - $443.77
Chemical Identifiers
CAS | 349-97-3 |
---|---|
Molecular Formula | C9H8F3NO |
Molecular Weight (g/mol) | 203.164 |
MDL Number | MFCD00013562 |
InChI Key | DFDHFECLWHHELH-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl acetanilide, n-4-trifluoromethyl phenyl acetamide, 4'-trifluoromethyl acetanilide, 4-trifluoromethylacetanilide, p-acetaminobenzotrifluoride, p-acetamidobenzotrifluoride, p-trifluoromethylacetanilide, p-acetylaminobenzotrifluoride, p-trifluoromethyl acetanilide |
PubChem CID | 67685 |
IUPAC Name | N-[4-(trifluoromethyl)phenyl]acetamide |
SMILES | CC(=O)NC1=CC=C(C=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1278906
|
Thermo Scientific Chemicals
A1278906 |
5 g |
Each for $145.36
|
|
|||||
AAA1278914
|
Thermo Scientific Chemicals
A1278914 |
25 g |
Each for $443.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
349-97-3 | |
203.164 | |
DFDHFECLWHHELH-UHFFFAOYSA-N | |
67685 | |
CC(=O)NC1=CC=C(C=C1)C(F)(F)F |
C9H8F3NO | |
MFCD00013562 | |
4-trifluoromethyl acetanilide, n-4-trifluoromethyl phenyl acetamide, 4'-trifluoromethyl acetanilide, 4-trifluoromethylacetanilide, p-acetaminobenzotrifluoride, p-acetamidobenzotrifluoride, p-trifluoromethylacetanilide, p-acetylaminobenzotrifluoride, p-trifluoromethyl acetanilide | |
N-[4-(trifluoromethyl)phenyl]acetamide |
Specifications
349-97-3 | |
Odorless | |
MFCD00013562 | |
2106965 | |
DFDHFECLWHHELH-UHFFFAOYSA-N | |
N-[4-(trifluoromethyl)phenyl]acetamide | |
67685 | |
≥98% |
150°C to 154°C | |
C9H8F3NO | |
5 g | |
4-trifluoromethyl acetanilide, n-4-trifluoromethyl phenyl acetamide, 4'-trifluoromethyl acetanilide, 4-trifluoromethylacetanilide, p-acetaminobenzotrifluoride, p-acetamidobenzotrifluoride, p-trifluoromethylacetanilide, p-acetylaminobenzotrifluoride, p-trifluoromethyl acetanilide | |
CC(=O)NC1=CC=C(C=C1)C(F)(F)F | |
203.164 | |
203.16 | |
4'-(Trifluoromethyl)acetanilide |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 206-495-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only