missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
4-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS
158063-66-2
Molecular Formula
C7H4F3NO2
Molecular Weight (g/mol)
191.11
MDL Number
MFCD00082626
InChI Key
LMRJHNFECNKDKH-UHFFFAOYSA-N
Synonym
4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
PubChem CID
2777549
IUPAC Name
4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES
OC(=O)C1=C(C=CN=C1)C(F)(F)F
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
RF03948CB
SDS
View Documents
Thermo Scientific Chemicals
RF03948CB
250mg
Amber Glass Bottle
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Specifications
Chemical Identifiers
CAS
158063-66-2
Molecular Weight (g/mol)
191.11
InChI Key
LMRJHNFECNKDKH-UHFFFAOYSA-N
PubChem CID
2777549
SMILES
OC(=O)C1=C(C=CN=C1)C(F)(F)F
Molecular Formula
C7H4F3NO2
MDL Number
MFCD00082626
Synonym
4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
IUPAC Name
4-(trifluoromethyl)pyridine-3-carboxylic acid
Specifications
CAS
158063-66-2
Assay Percent Range
97%
Molecular Formula
C7H4F3NO2
Quantity
250mg
InChI Key
LMRJHNFECNKDKH-UHFFFAOYSA-N
IUPAC Name
4-(trifluoromethyl)pyridine-3-carboxylic acid
PubChem CID
2777549
Percent Purity
97%
CAS Min %
97.0
Packaging
Amber Glass Bottle
MDL Number
MFCD00082626
Synonym
4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
SMILES
OC(=O)C1=C(C=CN=C1)C(F)(F)F
Molecular Weight (g/mol)
191.11
Formula Weight
191.11
Chemical Name or Material
4-(Trifluoromethyl)nicotinic acid
Videos
Safety and Handling
Safety and Handling
GHS H Statement: Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'