missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)phenylhydrazine hydrochloride, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3234003

 View more versions of this product

Catalog No. AAH3234003

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Trifluoromethyl)phenylhydrazine hydrochloride
∽210°C (decomposition)
MFCD00204233
Hygroscopic
WCAGNYIHAYOPSE-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]hydrazine;hydrochloride
24229790
96%
2923-56-0
C7H8ClF3N2
1 g
4-trifluoromethyl phenylhydrazine hydrochloride, 4-trifluoromethyl phenyl hydrazine hydrochloride, 1-4-trifluoromethyl phenyl hydrazine hydrochloride, 4-trifluoromethylphenyl hydrazine hydrochloride, 4-trifluoromethyl phenylhydrazine hcl, 4-trifluoromethyl phenylhydrazine, hcl, hydrazine, 4-trifluoromethyl phenyl-, monohydrochloride, n'-4-trifluoromethyl-phenyl-hydrazinium, chloride
C1=CC(=CC=C1C(F)(F)F)NN.Cl
212.6
212.6

Chemical Identifiers

2923-56-0
212.6
WCAGNYIHAYOPSE-UHFFFAOYSA-N
24229790
C1=CC(=CC=C1C(F)(F)F)NN.Cl
C7H8ClF3N2
MFCD00204233
4-trifluoromethyl phenylhydrazine hydrochloride, 4-trifluoromethyl phenyl hydrazine hydrochloride, 1-4-trifluoromethyl phenyl hydrazine hydrochloride, 4-trifluoromethylphenyl hydrazine hydrochloride, 4-trifluoromethyl phenylhydrazine hcl, 4-trifluoromethyl phenylhydrazine, hcl, hydrazine, 4-trifluoromethyl phenyl-, monohydrochloride, n'-4-trifluoromethyl-phenyl-hydrazinium, chloride
[4-(trifluoromethyl)phenyl]hydrazine;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 642-499-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only