Learn More
4-(Trifluoromethyl)phenylhydrazine hydrochloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3234009
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(Trifluoromethyl)phenylhydrazine hydrochloride | |
∽210°C (decomposition) | |
MFCD00204233 | |
Hygroscopic | |
WCAGNYIHAYOPSE-UHFFFAOYSA-N | |
[4-(trifluoromethyl)phenyl]hydrazine;hydrochloride | |
24229790 | |
96% |
2923-56-0 | |
C7H8ClF3N2 | |
10 g | |
4-trifluoromethyl phenylhydrazine hydrochloride, 4-trifluoromethyl phenyl hydrazine hydrochloride, 1-4-trifluoromethyl phenyl hydrazine hydrochloride, 4-trifluoromethylphenyl hydrazine hydrochloride, 4-trifluoromethyl phenylhydrazine hcl, 4-trifluoromethyl phenylhydrazine, hcl, hydrazine, 4-trifluoromethyl phenyl-, monohydrochloride, n'-4-trifluoromethyl-phenyl-hydrazinium, chloride | |
C1=CC(=CC=C1C(F)(F)F)NN.Cl | |
212.6 | |
212.6 |
Chemical Identifiers
2923-56-0 | |
212.6 | |
WCAGNYIHAYOPSE-UHFFFAOYSA-N | |
24229790 | |
C1=CC(=CC=C1C(F)(F)F)NN.Cl |
C7H8ClF3N2 | |
MFCD00204233 | |
4-trifluoromethyl phenylhydrazine hydrochloride, 4-trifluoromethyl phenyl hydrazine hydrochloride, 1-4-trifluoromethyl phenyl hydrazine hydrochloride, 4-trifluoromethylphenyl hydrazine hydrochloride, 4-trifluoromethyl phenylhydrazine hcl, 4-trifluoromethyl phenylhydrazine, hcl, hydrazine, 4-trifluoromethyl phenyl-, monohydrochloride, n'-4-trifluoromethyl-phenyl-hydrazinium, chloride | |
[4-(trifluoromethyl)phenyl]hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 642-499-3
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only