missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)thiophenol, 97%, Thermo Scientific Chemicals

$144.27 - $619.07

Chemical Identifiers

CAS 825-83-2
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00042320
InChI Key WCMLRSZJUIKVCW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
PubChem CID 136653
IUPAC Name 4-(trifluoromethyl)benzenethiol
SMILES C1=CC(=CC=C1C(F)(F)F)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1272503
View Documents
Thermo Scientific Chemicals
A1272503
1 g
Each for $144.27
 
AAA1272506
View Documents
Thermo Scientific Chemicals
A1272506
5 g
Each for $619.07
 
Description

Description

4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
Specifications

Chemical Identifiers

825-83-2
178.172
WCMLRSZJUIKVCW-UHFFFAOYSA-N
136653
C1=CC(=CC=C1C(F)(F)F)S
C7H5F3S
MFCD00042320
4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
4-(trifluoromethyl)benzenethiol

Specifications

825-83-2
66°C to 68°C (15 mmHg)
C7H5F3S
MFCD00042320
UN2810
Air Sensitive
Insoluble in water.
C1=CC(=CC=C1C(F)(F)F)S
178.172
178.18
4-(Trifluoromethyl)thiophenol
1.3
Stench
1.492
1 g
2045665
4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
WCMLRSZJUIKVCW-UHFFFAOYSA-N
4-(trifluoromethyl)benzenethiol
136653
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only