Learn More
4-Trifluoromethylphenylboronic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 359760050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Trifluoromethylphenylboronic acid | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00151855 | |
4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid | |
OB(O)C1=CC=C(C=C1)C(F)(F)F | |
189.93 | |
189.93 |
128796-39-4 | |
100.0 | |
97.5% min. (HPLC) | |
C7H6BF3O2 | |
5g | |
ALMFIOZYDASRRC-UHFFFAOYSA-N | |
[4-(trifluoromethyl)phenyl]boronic acid | |
2734389 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
RUO â Research Use Only