missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzylamine 97.0+%, TCI America™

$1261.97 - $1261.97

Chemical Identifiers

CAS 128273-56-3
Molecular Formula C8H8F3NS
Molecular Weight (g/mol) 207.214
MDL Number MFCD00190129
InChI Key LACURGWEZCFLBO-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzylamine, 4-trifluoromethyl thio phenyl methanamine, 4-trifluoromethylsulfanyl phenyl methanamine, 4-trifluoromethyl sulfanyl phenyl methanamine, 1-4-trifluoromethyl thio phenyl methanamine, 1-4-trifluoromethyl sulfanyl phenyl methanamine, acmc-1chvc, rarechem al bw 0450, 4-trifluomethylthio benzylamine
PubChem CID 2777875
IUPAC Name [4-(trifluoromethylsulfanyl)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)SC(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T27635G
View Documents
TCI America
T27635G
5 g
Each for $1,261.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

128273-56-3
207.214
LACURGWEZCFLBO-UHFFFAOYSA-N
2777875
C1=CC(=CC=C1CN)SC(F)(F)F
C8H8F3NS
MFCD00190129
4-trifluoromethylthio benzylamine, 4-trifluoromethyl thio phenyl methanamine, 4-trifluoromethylsulfanyl phenyl methanamine, 4-trifluoromethyl sulfanyl phenyl methanamine, 1-4-trifluoromethyl thio phenyl methanamine, 1-4-trifluoromethyl sulfanyl phenyl methanamine, acmc-1chvc, rarechem al bw 0450, 4-trifluomethylthio benzylamine
[4-(trifluoromethylsulfanyl)phenyl]methanamine

Specifications

128273-56-3
225°C
MFCD00190129
2735
LACURGWEZCFLBO-UHFFFAOYSA-N
[4-(trifluoromethylsulfanyl)phenyl]methanamine
2777875
≥97.0% (GC,T)
4-(Trifluoromethylthio)benzylamine
Colorless
C8H8F3NS
5 g
4-trifluoromethylthio benzylamine, 4-trifluoromethyl thio phenyl methanamine, 4-trifluoromethylsulfanyl phenyl methanamine, 4-trifluoromethyl sulfanyl phenyl methanamine, 1-4-trifluoromethyl thio phenyl methanamine, 1-4-trifluoromethyl sulfanyl phenyl methanamine, acmc-1chvc, rarechem al bw 0450, 4-trifluomethylthio benzylamine
C1=CC(=CC=C1CN)SC(F)(F)F
207.214
207.21
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents