missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Tritylaniline, 97%, Thermo Scientific Chemicals
$77.62 - $281.23
Chemical Identifiers
CAS | 22948-06-7 |
---|---|
Molecular Formula | C25H21N |
Molecular Weight (g/mol) | 335.45 |
MDL Number | MFCD00007898 |
InChI Key | XYHDHXBLSLSXSR-UHFFFAOYSA-N |
Synonym | p-tritylaniline, benzenamine, 4-triphenylmethyl, 4-triphenylmethyl aniline, p-aminotetraphenylmethane, 4-aminotetraphenylmethane, triphenyl p-aminophenyl methane, 4-trityl aniline, 4-triphenylmethyl benzenamine, p-toluidine, alpha,alpha,alpha-triphenyl, 4-aminophenyl triphenylmethane |
PubChem CID | 89914 |
IUPAC Name | 4-tritylaniline |
SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1055006
|
Thermo Scientific Chemicals
A1055006 |
5 g |
Each for $77.62
|
|
|||||
AAA1055014
|
Thermo Scientific Chemicals
A1055014 |
25 g |
Each for $281.23
|
|
|||||
Description
4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
22948-06-7 | |
335.45 | |
XYHDHXBLSLSXSR-UHFFFAOYSA-N | |
89914 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)N |
C25H21N | |
MFCD00007898 | |
p-tritylaniline, benzenamine, 4-triphenylmethyl, 4-triphenylmethyl aniline, p-aminotetraphenylmethane, 4-aminotetraphenylmethane, triphenyl p-aminophenyl methane, 4-trityl aniline, 4-triphenylmethyl benzenamine, p-toluidine, alpha,alpha,alpha-triphenyl, 4-aminophenyl triphenylmethane | |
4-tritylaniline |
Specifications
22948-06-7 | |
C25H21N | |
5 g | |
p-tritylaniline, benzenamine, 4-triphenylmethyl, 4-triphenylmethyl aniline, p-aminotetraphenylmethane, 4-aminotetraphenylmethane, triphenyl p-aminophenyl methane, 4-trityl aniline, 4-triphenylmethyl benzenamine, p-toluidine, alpha,alpha,alpha-triphenyl, 4-aminophenyl triphenylmethane | |
XYHDHXBLSLSXSR-UHFFFAOYSA-N | |
4-tritylaniline | |
89914 | |
97% |
256°C to 260°C | |
MFCD00007898 | |
2750328 | |
Insoluble in water. | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)N | |
335.45 | |
335.45 | |
4-Tritylaniline |
Safety and Handling
EINECSNumber : 245-346-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only