missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Vinylbenzylamine (stabilized with MEHQ) 92.0+%, TCI America™
$257.24 - $877.97
Chemical Identifiers
| CAS | 50325-49-0 |
|---|---|
| Molecular Formula | C9H11N |
| Molecular Weight (g/mol) | 133.194 |
| MDL Number | MFCD08704374 |
| InChI Key | WAVDSLLYAQBITE-UHFFFAOYSA-N |
| Synonym | 4-(Aminomethyl)styrene |
| PubChem CID | 11073466 |
| IUPAC Name | (4-ethenylphenyl)methanamine |
| SMILES | C=CC1=CC=C(C=C1)CN |
Chemical Identifiers
| 50325-49-0 | |
| 133.194 | |
| WAVDSLLYAQBITE-UHFFFAOYSA-N | |
| 11073466 | |
| C=CC1=CC=C(C=C1)CN |
| C9H11N | |
| MFCD08704374 | |
| 4-(Aminomethyl)styrene | |
| (4-ethenylphenyl)methanamine |
Specifications
| 50325-49-0 | |
| 60°C | |
| MFCD08704374 | |
| 4-(Aminomethyl)styrene | |
| C=CC1=CC=C(C=C1)CN | |
| 133.194 | |
| 133.19 | |
| Liquid |
| Yellow | |
| C9H11N | |
| 1 g | |
| WAVDSLLYAQBITE-UHFFFAOYSA-N | |
| (4-ethenylphenyl)methanamine | |
| 11073466 | |
| ≥92.0% (GC) | |
| 4-Vinylbenzylamine (stabilized with MEHQ) |
Safety and Handling
TSCA : No
Recommended Storage : Freezer