Learn More
4-Vinylphenol, 95%, 10% solution in propylene glycol, Thermo Scientific Chemicals
$548.62 - $548.62
Chemical Identifiers
| CAS | 2628-17-3 |
|---|---|
| Molecular Formula | C8H8O |
| Molecular Weight (g/mol) | 120.16 |
| InChI Key | FUGYGGDSWSUORM-UHFFFAOYSA-N |
| Synonym | 4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene |
| PubChem CID | 62453 |
| ChEBI | CHEBI:1883 |
| IUPAC Name | 4-ethenylphenol |
| SMILES | C=CC1=CC=C(C=C1)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC434040250
|
Thermo Scientific Chemicals
434040250 |
25 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2628-17-3 | |
| 120.16 | |
| 4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene | |
| CHEBI:1883 | |
| C=CC1=CC=C(C=C1)O |
| C8H8O | |
| FUGYGGDSWSUORM-UHFFFAOYSA-N | |
| 62453 | |
| 4-ethenylphenol |
Specifications
| 2628-17-3,57-55-6 | |
| 91.0,11.0 | |
| 1.0390g/mL | |
| Authentic | |
| Glass bottle | |
| 1.4420 to 1.4520 (20°C, 589nm) | |
| 1.039 | |
| FUGYGGDSWSUORM-UHFFFAOYSA-N | |
| 4-ethenylphenol | |
| 62453 | |
| 120.16 | |
| Liquid |
| 89.0,9.0 | |
| Colorless to Orange | |
| 189°C | |
| 9 to 15% (GC) | |
| C8H8O | |
| 25 mL | |
| 4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene | |
| C=CC1=CC=C(C=C1)O | |
| 120.16 | |
| CHEBI:1883 | |
| 95% | |
| 4-Vinylphenol |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 220-103-6
RUO – Research Use Only