missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylphenol, min 10% soln. in propylene glycol

CAS: 2628-17-3 | C8H8O | 120.151 g/mol

$94.97 - $289.51

Chemical Identifiers

CAS 2628-17-3
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00017593
InChI Key FUGYGGDSWSUORM-UHFFFAOYSA-N
Synonym 4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
PubChem CID 62453
ChEBI CHEBI:1883
IUPAC Name 4-ethenylphenol
SMILES C=CC1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1090203
View Documents
Thermo Scientific Chemicals
L1090203
1 g N/A
Only null left
 
AAL1090206
View Documents
Thermo Scientific Chemicals
L1090206
5 g N/A
Only null left
 
Description

Description

4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Moisture and heat sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2628-17-3
120.151
FUGYGGDSWSUORM-UHFFFAOYSA-N
62453
4-ethenylphenol
C8H8O
MFCD00017593
4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
CHEBI:1883
C=CC1=CC=C(C=C1)O

Specifications

2628-17-3
>100°C (212°F)
1.447
1 g
Hygroscopic
Slightly miscible with water.
C=CC1=CC=C(C=C1)O
120.151
CHEBI:1883
≥10% soln. in propylene glycol
1.04
C8H8O
MFCD00017593
506844
4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
FUGYGGDSWSUORM-UHFFFAOYSA-N
4-ethenylphenol
62453
120.15
4-Vinylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P201-P202-P260-P264b-P270-P281-P308+P313-P501c

H341-H361-H372

EINECSNumber : 220-103-6

RTECSNumber : SN3800000

TSCA : No

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only