Thermo Scientific™ 4-Way Flipper™ Racks
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Supplier: Thermo Scientific™ 8850
Description
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.
Specifications
4-way Flipper Rack | |
Fluorescent Green | |
Thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side | |
20 Racks |
4-Way Flipper Rack | |
0.5 mL, 15 mL, Tubes (50 mL) | |
Rectangular |