Learn More
Agilent BioTek 405 TS Touch Microplate Washer, IVD
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek 405TSRSQSI
Description
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.
The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Specifications
ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes | |
USB (2) | |
<3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) | |
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates | |
96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s | |
Microplate Washer | |
1 to 250 | |
LED | |
Internal positive displacement pump | |
50/60 Hz | |
Ultrasonic Advantage; 96-well microplate washing; Verify Technology | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
With internal buffer switching - 36 lb.; Without - 30 lb. |
CE and TUV marked. RoHS compliant. | |
Optional (4 or 10 L) | |
25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) | |
≤2 μL/well | |
Programmable in minutes and seconds, up to 60 min. | |
With internal buffer switching - 16.5 kg; Without - 13.5 kg | |
25 to 3000 μL/well | |
High to Low Flow; Optimized rates for cell assays | |
ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays | |
405TSRSQ | |
14 in. | |
10 in. | |
17 in. | |
High Resolution LED Backlit Touch Screen | |
100/240 V |