Cytiva HyCell™ TransFx™ H Media
Supplier: Cytiva SH3093902
Description
HyCell TransFx-H media for transient transfection of HEK293 cells and production of a variety of recombinant proteins manufactured under cGMP in ISO-compliant facilities.
Highlights:
- Defined hydrolysate-free ADCF cell culture media formulation
- Supports transient transfection and growth of multiple HEK293 cell lines
- Supports the transient expression of a variety of recombinant proteins
- Same media for transient transfection and production
- Developed through our Metabolic Pathway Design process for high transfection efficiency, high viable cell yields, and recombinant protein production
- Component traceability
- Manufactured under cGMP in ISO-compliant facility
Specifications
2° to 8°C, away from light | |
HyCell TransFx-H liquid medium - 1000 mL | |
1000 mL |
Liquid | |
0.2 μm Sterile-filtered |