missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer
Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.
$145.93 - $233.00
Specifications
Bottle Size | 2.5 x 6.3 cm (1 x 2.5 in.) |
---|---|
For Use With (Application) | Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators |
Resolution | 0.1° |
Weight (English) | 4 oz. |
Product Type | Refrigerator/Freezer Ultra Thermometer |
Includes: Flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
Catalog Number | Mfr. No. | Probe Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Probe Type | Price | Quantity | |||||
14-650-103
|
Fisherbrand™
9000936 |
1 Bottle Probe |
Each for $183.03
|
|
|||||
14-650-104
|
Fisherbrand™
9367787 |
Two Bottle Probes |
Each for $219.38
|
|
|||||
14-650-105
|
Fisherbrand™
9367793 |
One Bullet Probe |
Each for $172.38
|
|
|||||
14-650-106
|
Fisherbrand™
9367798 |
Two Bullet Probes |
Each for $177.29
|
|
|||||
14-650-107
|
Fisherbrand™
9367802 |
1 Bottle and 1 Bullet Probe |
Each for $190.56
|
|
|||||
14-650-108
|
Fisherbrand™
9367806 |
1 x 5 mL Vaccine Bottle Probe |
Each for $186.79
|
|
|||||
14-650-109
|
Fisherbrand™
9367811 |
2 x 5 mL Vaccine Bottle Probe |
Each for $228.53
|
|
|||||
15-079-659
|
Fisherbrand™
9367818 |
2 Plastic Bottle Probes |
Each for $233.00
|
|
|||||
36-104-1622
|
N/A | 1 Plastic Bottle Probe |
Each for $145.93
|
|
|||||
Description
Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.
- Designed to meet current CDC requirements for vaccine storage and monitoring
- Triple display simultaneously shows min., max. and current temperatures for a specified probe
- Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
- Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
- °C/°F switchable
- Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
- Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
- Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
- Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
- Ten-foot micro-cable permits refrigerator doors to close on it
- Range: -50° to +70°C (-58° to +158°F)
- Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
- Resolution: 0.1°
Certifications
Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.
Specifications
2.5 x 6.3 cm (1 x 2.5 in.) | |
0.1° | |
Refrigerator/Freezer Ultra Thermometer | |
70°C | |
-50°C | |
-50°C to +70°C | |
Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator | |
Digital |
Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators | |
4 oz. | |
158°F | |
-58°F | |
-58°F to +158°F | |
Yes | |
±0.5°C (-30°C to +50°C), ±1°C otherwise |