missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 24-well Standard Plate
Sterile and individually wrapped and available with either untreated or cell culture treated surface.
$714.25 - $759.25
Specifications
| Closure Included | Yes |
|---|---|
| Bottom Shape | Flat |
| Capacity (Metric) | 3.1 mL |
| Color | Transparent |
| Culture Area | 210 mm2 |
| Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity | |||||
|
14-380-865
|
BrandTech™ BRAND™
782880 |
pureGrade S |
Pack of 10 Each for $714.25
|
|
|||||
|
14-380-867
|
BrandTech™ BRAND™
782890 |
cellGrade plus |
Pack of 10 Each for $759.25
|
|
|||||
Description
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Available with pureGrade™ S and cellGrade™ plus surfaces
- Includes 24 individually fillable wells that can be fitted with strips of 4 inserts and/or individual inserts
- Guide ridges for perfect insert fit
- Designated feeding ports for easy media changes, suitable for robotics/automation
- 10 plates per pack (individually wrapped with lid)
Surface Information:
- pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
- cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.
Applications:
- Ideal for co-culture, secretions studies, and migrations studies
Compliance
ANSI/SLAS standards 1 and 4
Specifications
| Yes | |
| 3.1 mL | |
| 210 mm2 | |
| Sterile | |
| 24 | |
| Insert system |
| Flat | |
| Transparent | |
| Polystyrene | |
| Cell Culture Insert System | |
| Co-culture, secretion studies, migration studies | |
| 19.5 mm |