missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Excelta™ Serrated Locking Hemostat

Available in Straight/Curved shape

Supplier:  Excelta™ 40SE

 View more versions of this product

Catalog No. 18-100-969


Explore available promotions

Description

Description

  • Clean room safe
  • Made of stainless steel
  • Overall length: 200mm (8 in.)

Specifications

Specifications

Locking hemostat
200 mm
Curved
8 in.
Stainless Steel
Videos
SDS
Documents

Documents

Promotions

Promotions