missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5,10,15,20-Tetra(4-pyridyl)porphyrin 96.0+%, TCI America™
Supplier: TCI America T22225G
Specifications
5,10,15,20-Tetra(4-pyridyl)porphyrin | |
Black-Purple | |
MFCD00009626 | |
5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine | |
N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1 | |
618.70 | |
618.70 | |
Crystalline Powder |
16834-13-2 | |
C40H26N8 | |
5 g | |
WQCKXOJXOKSXQZ-UHFFFAOYSA-N | |
2,7,12,17-tetrakis(pyridin-4-yl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,â¶.1â¸,¹¹.1¹³,¹â¶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene | |
86073 | |
≥96.0% (N) |
Chemical Identifiers
16834-13-2 | |
618.70 | |
WQCKXOJXOKSXQZ-UHFFFAOYSA-N | |
86073 | |
N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1 |
C40H26N8 | |
MFCD00009626 | |
5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine | |
2,7,12,17-tetrakis(pyridin-4-yl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,â¶.1â¸,¹¹.1¹³,¹â¶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene |
Safety and Handling
TSCA : Yes