missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin 90.0+%, TCI America™

Supplier:  TCI America T18151G

 View more versions of this product

Catalog No. T18151G


Explore available promotions

Description

Description

Specifications

Specifications

5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin
Blue-Purple
MFCD02093497
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)-21H,23H-porphine
C1=CC2=C(C3=CC=C(N3)C(=C4C=CC(=N4)C(=C5C=CC(=N5)C(=C1N2)C6=CC(=CC(=C6)O)O)C7=CC(=CC(=C7)O)O)C8=CC(=CC(=C8)O)O)C9=CC(=CC(=C9)O)O
742.744
742.74
Crystalline Powder
145764-54-1
C44H30N4O8
1 g
XDVQXKKXBXLGCO-UHFFFAOYSA-N
5-[10,15,20-tris(3,5-dihydroxyphenyl)-21,24-dihydroporphyrin-5-yl]benzene-1,3-diol
471968
≥90.0% (HPLC)

Chemical Identifiers

145764-54-1
742.744
XDVQXKKXBXLGCO-UHFFFAOYSA-N
471968
C1=CC2=C(C3=CC=C(N3)C(=C4C=CC(=N4)C(=C5C=CC(=N5)C(=C1N2)C6=CC(=CC(=C6)O)O)C7=CC(=CC(=C7)O)O)C8=CC(=CC(=C8)O)O)C9=CC(=CC(=C9)O)O
C44H30N4O8
MFCD02093497
5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)-21H,23H-porphine
5-[10,15,20-tris(3,5-dihydroxyphenyl)-21,24-dihydroporphyrin-5-yl]benzene-1,3-diol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions