missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone 98.0+%, TCI America™
Supplier: TCI America E11345G
Specifications
5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone | |
145°C | |
MFCD03093929 | |
MXQUEDUMKWBYHI-UHFFFAOYSA-N | |
5-(2-ethoxyphenyl)-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one | |
899745 | |
≥98.0% (HPLC,T) |
139756-21-1 | |
C17H20N4O2 | |
5 g | |
CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC=CC=C1OCC | |
312.37 | |
312.37 | |
Crystalline Powder |
Chemical Identifiers
139756-21-1 | |
312.37 | |
MXQUEDUMKWBYHI-UHFFFAOYSA-N | |
5-(2-ethoxyphenyl)-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one |
C17H20N4O2 | |
MFCD03093929 | |
899745 | |
CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC=CC=C1OCC |
Safety and Handling
TSCA : No