missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC46901DA

 View more versions of this product

Catalog No. CC46901DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carboxylic acid
96.5
97%
C9H8N2O3
1g
XXEWHNSDJMBLBK-UHFFFAOYSA-N
5-(furan-2-yl)-1-methyl-1H-pyrazole-3-carboxylic acid
7537639
97%
108128-39-8
100.0
Amber Glass Bottle
MFCD08060535
5-2-furyl-1-methyl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-1-methylpyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-1-methyl, 5-furan-2-yl-1-methyl-1h-pyrazole-3-carboxylic acid, acmc-1btkv, 5-2-furyl-1-methylpyrazole-3-carboxylic acid
CN1N=C(C=C1C1=CC=CO1)C(O)=O
192.17
192.17

Chemical Identifiers

108128-39-8
192.17
XXEWHNSDJMBLBK-UHFFFAOYSA-N
7537639
CN1N=C(C=C1C1=CC=CO1)C(O)=O
C9H8N2O3
MFCD08060535
5-2-furyl-1-methyl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-1-methylpyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl-1-methyl, 5-furan-2-yl-1-methyl-1h-pyrazole-3-carboxylic acid, acmc-1btkv, 5-2-furyl-1-methylpyrazole-3-carboxylic acid
5-(furan-2-yl)-1-methyl-1H-pyrazole-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions