missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Methyl-1,3-thiazol-4-yl)-3-isoxazolecarboxylic acid, 97%, Thermo Scientific™

$283.10 - $283.10

Chemical Identifiers

CAS 368870-05-7
Molecular Formula C8H6N2O3S
Molecular Weight (g/mol) 210.207
MDL Number MFCD03086107
InChI Key QAXQRHWAJNDTCV-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-3-isoxazolecarboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-1,2-oxazole-3-carboxylic acid, 3-isoxazolecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-3-isoxazolecarboxyli, 5-2-methylthiazol-4-yl isoxazole-3-carboxylic acid, 3-carboxy-5-2-methyl-1,3-thiazol-4-yl-1,2-oxazole, 4-3-carboxy-1,2-oxazol-5-yl-2-methyl-1,3-thiazole, 5-2-methyl-1,3-thiazol-4-yl isoxazole-3-carboxylic acid
PubChem CID 2776408
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazole-3-carboxylic acid
SMILES CC1=NC(=CS1)C2=CC(=NO2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC14001CB
SDS View Documents
Thermo Scientific Chemicals
CC14001CB
250 mg Amber Glass Bottle N/A
 
Description

Description

Specifications

Chemical Identifiers

368870-05-7
210.207
QAXQRHWAJNDTCV-UHFFFAOYSA-N
2776408
CC1=NC(=CS1)C2=CC(=NO2)C(=O)O
C8H6N2O3S
MFCD03086107
5-2-methyl-1,3-thiazol-4-yl-3-isoxazolecarboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-1,2-oxazole-3-carboxylic acid, 3-isoxazolecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-3-isoxazolecarboxyli, 5-2-methylthiazol-4-yl isoxazole-3-carboxylic acid, 3-carboxy-5-2-methyl-1,3-thiazol-4-yl-1,2-oxazole, 4-3-carboxy-1,2-oxazol-5-yl-2-methyl-1,3-thiazole, 5-2-methyl-1,3-thiazol-4-yl isoxazole-3-carboxylic acid
5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazole-3-carboxylic acid

Specifications

368870-05-7
100.0
Amber Glass Bottle
MFCD03086107
5-2-methyl-1,3-thiazol-4-yl-3-isoxazolecarboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-1,2-oxazole-3-carboxylic acid, 3-isoxazolecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-3-isoxazolecarboxyli, 5-2-methylthiazol-4-yl isoxazole-3-carboxylic acid, 3-carboxy-5-2-methyl-1,3-thiazol-4-yl-1,2-oxazole, 4-3-carboxy-1,2-oxazol-5-yl-2-methyl-1,3-thiazole, 5-2-methyl-1,3-thiazol-4-yl isoxazole-3-carboxylic acid
CC1=NC(=CS1)C2=CC(=NO2)C(=O)O
210.207
210.21
5-(2-Methyl-1, 3-thiazol-4-yl)-3-isoxazolecarboxylic acid
96.5
97%
C8H6N2O3S
250 mg
QAXQRHWAJNDTCV-UHFFFAOYSA-N
5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazole-3-carboxylic acid
2776408
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'