missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Pyridyl)-1,3-oxazole, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H34123MD

 View more versions of this product

Catalog No. AAH34123MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-(2-Pyridyl)-1,3-oxazole
C8H6N2O
250 mg
LUMKXMDGONYZHO-UHFFFAOYSA-N
5-pyridin-2-yl-1,3-oxazole
737194
97%
70380-73-3
MFCD00085160
5-2-pyridyl-1,3-oxazole, 2-1,3-oxazol-5-yl pyridine, 5-pyridin-2-yl oxazole, pyridine,2-5-oxazolyl, 2-5-oxazolyl pyridine, 2-oxazol-5-yl-pyridine, 5-pyridin-2-yl-1,3-oxazole, pyridine, 2-5-oxazolyl, 5-2-pyridyl oxazole, acmc-209oeb
C1=CC=NC(=C1)C2=CN=CO2
146.149
146.15

Chemical Identifiers

70380-73-3
146.149
LUMKXMDGONYZHO-UHFFFAOYSA-N
5-pyridin-2-yl-1,3-oxazole
C8H6N2O
MFCD00085160
737194
C1=CC=NC(=C1)C2=CN=CO2
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only