missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Pyridyl)thiophene-2-sulfonyl chloride, Technical Grade, Thermo Scientific™

Chemical Identifiers

CAS 151858-64-9
Molecular Formula C9H6ClNO2S2
Molecular Weight (g/mol) 259.72
MDL Number MFCD00052284
InChI Key OGOMWPWAEIDEOU-UHFFFAOYSA-N
Synonym 5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride
PubChem CID 2737231
IUPAC Name 5-pyridin-2-ylthiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC02203DA
SDS View Documents
Thermo Scientific Chemicals
CC02203DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC02203DE
SDS View Documents
Thermo Scientific Chemicals
CC02203DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

151858-64-9
259.72
OGOMWPWAEIDEOU-UHFFFAOYSA-N
2737231
ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1
C9H6ClNO2S2
MFCD00052284
5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride
5-pyridin-2-ylthiophene-2-sulfonyl chloride

Specifications

151858-64-9
C9H6ClNO2S2
1g
OGOMWPWAEIDEOU-UHFFFAOYSA-N
5-pyridin-2-ylthiophene-2-sulfonyl chloride
2737231
95%
95%
MFCD00052284
5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride
ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1
259.72
259.74
5-(2-Pyridyl)thiophene-2-sulfonyl chloride
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'