missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%, Thermo Scientific Chemicals

A tricyclic serotnonin reuptake inhibitor

$266.62 - $712.67

Chemical Identifiers

CAS 549-18-8
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.869
MDL Number MFCD00012537
InChI Key KFYRPLNVJVHZGT-UHFFFAOYSA-N
Synonym amitriptyline hydrochloride, amitriptyline hcl, annoyltin, tryptizol, domical, elavil hydrochloride, lentizol, saroten, amavil, ami-anelun
PubChem CID 11065
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b
SMILES CN(C)CCC=C1C2=CC=CC=C2CCC3=CC=CC=C31.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6559614
View Documents
Thermo Scientific Chemicals
J6559614
25 g
Each for $266.62
 
AAJ6559622
View Documents
Thermo Scientific Chemicals
J6559622
100 g
Each for $712.67
 
Description

Description

A tricyclic serotnonin reuptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

549-18-8
313.869
KFYRPLNVJVHZGT-UHFFFAOYSA-N
11065
CN(C)CCC=C1C2=CC=CC=C2CCC3=CC=CC=C31.Cl
C20H24ClN
MFCD00012537
amitriptyline hydrochloride, amitriptyline hcl, annoyltin, tryptizol, domical, elavil hydrochloride, lentizol, saroten, amavil, ami-anelun
3-(5,6-dihydrodibenzo[2,1-b

Specifications

549-18-8
White
MFCD00012537
UN2811
amitriptyline hydrochloride, amitriptyline hcl, annoyltin, tryptizol, domical, elavil hydrochloride, lentizol, saroten, amavil, ami-anelun
CN(C)CCC=C1C2=CC=CC=C2CCC3=CC=CC=C31.Cl
313.869
313.86
Powder
196°C to 197°C
C20H24ClN
25 g
14,487
KFYRPLNVJVHZGT-UHFFFAOYSA-N
3-(5,6-dihydrodibenzo[2,1-b
11065
98%
5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H334-H361-H315-H319-H317-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P281-P301+P310-P307+P311-P330-P501c

H301-H361d-H370

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 208-964-6

RTECSNumber : HO9450000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only