missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(4-Chlorophenyl)furfural, 95+%, Thermo Scientific™

Chemical Identifiers

CAS 34035-03-5
Molecular Formula C11H7ClO2
Molecular Weight (g/mol) 206.63
MDL Number MFCD00195947
InChI Key ROJGJNINTRCMBL-UHFFFAOYSA-N
Synonym 5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
PubChem CID 676184
IUPAC Name 5-(4-chlorophenyl)furan-2-carbaldehyde
SMILES ClC1=CC=C(C=C1)C1=CC=C(O1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC375500010
SDS View Documents
Thermo Scientific Chemicals
375500010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC375500050
SDS View Documents
Thermo Scientific Chemicals
375500050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

34035-03-5
206.63
ROJGJNINTRCMBL-UHFFFAOYSA-N
676184
ClC1=CC=C(C=C1)C1=CC=C(O1)C=O
C11H7ClO2
MFCD00195947
5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
5-(4-chlorophenyl)furan-2-carbaldehyde

Specifications

34035-03-5
100.0
95% min. (GC)
C11H7ClO2
1g
ROJGJNINTRCMBL-UHFFFAOYSA-N
5-(4-chlorophenyl)furan-2-carbaldehyde
676184
95+%
95.0
Authentic
Glass bottle
MFCD00195947
5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
ClC1=CC=C(C=C1)C1=CC=C(O1)C=O
206.63
206.63
5-(4-Chlorophenyl)furfural
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only