missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxyphenyl)hydantoin 98.0+%, TCI America™

$65.24 - $192.00

Chemical Identifiers

CAS 2420-17-9
Molecular Formula C9H8N2O3
Molecular Weight (g/mol) 192.17
MDL Number MFCD00044002
InChI Key UMTNMIARZPDSDI-UHFFFAOYNA-N
PubChem CID 94233
IUPAC Name 5-(4-hydroxyphenyl)imidazolidine-2,4-dione
SMILES OC1=CC=C(C=C1)C1NC(=O)NC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08065G
View Documents
TCI America
H08065G
5 g
Each for $65.24
Only null left
 
H080625G
View Documents
TCI America
H080625G
25 g
Each for $192.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2420-17-9
192.17
UMTNMIARZPDSDI-UHFFFAOYNA-N
5-(4-hydroxyphenyl)imidazolidine-2,4-dione
C9H8N2O3
MFCD00044002
94233
OC1=CC=C(C=C1)C1NC(=O)NC1=O

Specifications

2420-17-9
C9H8N2O3
5 g
OC1=CC=C(C=C1)C1NC(=O)NC1=O
192.17
192.17
Crystalline Powder
White-Yellow
MFCD00044002
UMTNMIARZPDSDI-UHFFFAOYNA-N
5-(4-hydroxyphenyl)imidazolidine-2,4-dione
94233
≥98.0% (N)
5-(4-Hydroxyphenyl)hydantoin
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5364

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'