missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(4-Morpholinyl)-2-nitrophenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2495006

 View more versions of this product

Catalog No. AAB2495006


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-(4-Morpholinyl)-2-nitrophenol
143°C to 145°C
MFCD00053057
5-morpholino-2-nitrophenol, 5-4-morpholinyl-2-nitrophenol, 5-4-morpholino-2-nitrophenol, 5-morpholin-4-yl-2-nitrophenol, 5-morpholin-4-yl-2-nitro-phenolate, maybridge1_001296, acmc-1c6gm, 2-nitro-5-morpholinophenol, 5-morpholin-4-yl-2-nitro-phenol, n-3-hydroxy-4-nitrophenyl morpholine
C1COCCN1C2=CC(=C(C=C2)[N+](=O)[O-])O
224.216
224.22
175135-19-0
C10H12N2O4
5 g
XXIIDARBNMZFEH-UHFFFAOYSA-N
5-morpholin-4-yl-2-nitrophenol
2799399
97%

Chemical Identifiers

175135-19-0
224.216
XXIIDARBNMZFEH-UHFFFAOYSA-N
2799399
C1COCCN1C2=CC(=C(C=C2)[N+](=O)[O-])O
C10H12N2O4
MFCD00053057
5-morpholino-2-nitrophenol, 5-4-morpholinyl-2-nitrophenol, 5-4-morpholino-2-nitrophenol, 5-morpholin-4-yl-2-nitrophenol, 5-morpholin-4-yl-2-nitro-phenolate, maybridge1_001296, acmc-1c6gm, 2-nitro-5-morpholinophenol, 5-morpholin-4-yl-2-nitro-phenol, n-3-hydroxy-4-nitrophenyl morpholine
5-morpholin-4-yl-2-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only