missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6,7,8-Tetrahydroquinoxaline, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1120406

 View more versions of this product

Catalog No. AAL1120406


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5,6,7,8-Tetrahydroquinoxaline
1.061
92°C (197°F)
1.542
5 g
cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495
C1CCC2=NC=CN=C2C1
134.182
134.18
34413-35-9
72°C to 74°C (4 mmHg)
C8H10N2
MFCD00038078
957053
XCZPDOCRSYZOBI-UHFFFAOYSA-N
5,6,7,8-tetrahydroquinoxaline
36822
99%

Chemical Identifiers

34413-35-9
134.182
XCZPDOCRSYZOBI-UHFFFAOYSA-N
36822
C1CCC2=NC=CN=C2C1
C8H10N2
MFCD00038078
cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495
5,6,7,8-tetrahydroquinoxaline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 252-002-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only