missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 5(6)-Carboxyfluorescein

$53.04 - $266.15

Chemical Identifiers

CAS 72088-94-9
Molecular Formula C42H24O14
Molecular Weight (g/mol) 752.64
MDL Number MFCD00151081
InChI Key BPVHBBXCESDRKW-UHFFFAOYSA-N
Synonym 5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid
PubChem CID 44119975
IUPAC Name 3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC404105000
SDS View Documents
Thermo Scientific Chemicals
404105000
500 mg Glass bottle
Each for $53.04
 
AC404100050
SDS View Documents
Thermo Scientific Chemicals
404100050
5 g Glass bottle
Each for $266.15
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

72088-94-9
752.64
BPVHBBXCESDRKW-UHFFFAOYSA-N
44119975
OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2
C42H24O14
MFCD00151081
5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid
3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid

Specifications

72088-94-9
99%
C42H24O14
500 mg
Solubility in water: slightly soluble. Other solubilities: soluble in 0.1 n naoh
OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2
752.64
376.32
275°C
Glass bottle
MFCD00151081
5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid
BPVHBBXCESDRKW-UHFFFAOYSA-N
3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
44119975
5(6)-Carboxyfluorescein, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 276-331-1

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only