missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dibromo-1-hexene 96.0+%, TCI America™

$203.17 - $704.62

Chemical Identifiers

CAS 4285-48-7
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.954
MDL Number MFCD00041698
InChI Key WFGDPUOTONXJOS-UHFFFAOYSA-N
PubChem CID 2755398
IUPAC Name 5,6-dibromohex-1-ene
SMILES C=CCCC(CBr)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D48441G
View Documents
TCI America
D48441G
1 g
Each for $203.17
Only null left
 
D48445G
missing translation for 'viewDocuments'
TCI America
D48445G
5 g
Each for $704.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4285-48-7
241.954
WFGDPUOTONXJOS-UHFFFAOYSA-N
5,6-dibromohex-1-ene
C6H10Br2
MFCD00041698
2755398
C=CCCC(CBr)Br

Specifications

4285-48-7
55°C
MFCD00041698
WFGDPUOTONXJOS-UHFFFAOYSA-N
5,6-dibromohex-1-ene
2755398
≥96.0% (GC)
5,6-Dibromo-1-hexene
Yellow
C6H10Br2
1 g
C=CCCC(CBr)Br
241.954
241.95
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents