missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, 97+%, Thermo Scientific Chemicals

$72.67 - $195.40

Chemical Identifiers

CAS 1264198-47-1
Molecular Formula C20H12N4Na2O6S2
Molecular Weight (g/mol) 514.44
MDL Number MFCD00717621
InChI Key DDIHLFCSEHAOJZ-UHFFFAOYSA-L
Synonym ferrozin, ferrozine, ferrozine disodium salt, unii-29j2ciw9pk, 29j2ciw9pk, 3-2-pyridyl-5,6-di 4-phenylsulfonic acid-1,2,4-triazine, disodium salt, 4,4'-3-2-pyridyl-triazine-5,6-diyl dibenzenesulfonic acid disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid, disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid,disodium salt, benzenesulfonic acid, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-, disodium salt
PubChem CID 34127
IUPAC Name disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1060703
View Documents
Thermo Scientific Chemicals
L1060703
1 g
Each for $72.67
 
AAL1060706
View Documents
Thermo Scientific Chemicals
L1060706
5 g
Each for $195.40
 
Description

Description

Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

1264198-47-1
514.44
DDIHLFCSEHAOJZ-UHFFFAOYSA-L
34127
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1
C20H12N4Na2O6S2
MFCD00717621
ferrozin, ferrozine, ferrozine disodium salt, unii-29j2ciw9pk, 29j2ciw9pk, 3-2-pyridyl-5,6-di 4-phenylsulfonic acid-1,2,4-triazine, disodium salt, 4,4'-3-2-pyridyl-triazine-5,6-diyl dibenzenesulfonic acid disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid, disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid,disodium salt, benzenesulfonic acid, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-, disodium salt
disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate

Specifications

1264198-47-1
MFCD00717621
5222087
Soluble in water.
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1
514.44
514.46 (Anhydrous)
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate
C20H12N4Na2O6S2
1 g
ferrozin, ferrozine, ferrozine disodium salt, unii-29j2ciw9pk, 29j2ciw9pk, 3-2-pyridyl-5,6-di 4-phenylsulfonic acid-1,2,4-triazine, disodium salt, 4,4'-3-2-pyridyl-triazine-5,6-diyl dibenzenesulfonic acid disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid, disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid,disodium salt, benzenesulfonic acid, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-, disodium salt
DDIHLFCSEHAOJZ-UHFFFAOYSA-L
disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
34127
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 248-797-6

RTECSNumber : DB7345000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only