Learn More
5,7-Dihydroxy-4'-methoxyisoflavone, 98%, Thermo Scientific Chemicals
$141.70 - $1694.70
Chemical Identifiers
CAS | 491-80-5 |
---|---|
Molecular Formula | C16H12O5 |
Molecular Weight (g/mol) | 284.27 |
MDL Number | MFCD00006839 |
InChI Key | WUADCCWRTIWANL-UHFFFAOYSA-N |
Synonym | biochanin a, biochanin, 4'-methylgenistein, 5,7-dihydroxy-4'-methoxyisoflavone, 5,7-dihydroxy-3-4-methoxyphenyl-4h-chromen-4-one, genistein 4-methyl ether, pratensol, biochanine a, 5,7-dihydrox-4'-methoxyisoflavone, 4h-1-benzopyran-4-one, 5,7-dihydroxy-3-4-methoxyphenyl |
PubChem CID | 5280373 |
ChEBI | CHEBI:17574 |
IUPAC Name | 5,7-dihydroxy-3-(4-methoxyphenyl)chromen-4-one |
SMILES | COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA43853MD
|
Thermo Scientific Chemicals
043853MD |
250 mg |
Each for $141.70
|
|
||||||
AA4385303
|
Thermo Scientific Chemicals
04385303 |
1 g |
Each for $401.75
|
|
||||||
AA4385306
|
Thermo Scientific Chemicals
04385306 |
5 g |
Each for $1,694.70
|
|
||||||
Description
It has putative benefits in dietary cancer prophylaxis. It has also been found to inhibit fatty acid amide hydrolase and to act as agonist of PPARgamma, nuclear receptor that is current pharmacological target for the treatment of diabetes type 2. It acts as an antineoplastic agent. It is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels. Induction of sulfotransferases for xenobiotic detoxification has been proposed as a mechanism of its cancer preventive effects. It is a nitric oxide synthase inhibitor and apoptosis inducer
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
491-80-5 | |
284.27 | |
WUADCCWRTIWANL-UHFFFAOYSA-N | |
5280373 | |
5,7-dihydroxy-3-(4-methoxyphenyl)chromen-4-one |
C16H12O5 | |
MFCD00006839 | |
biochanin a, biochanin, 4'-methylgenistein, 5,7-dihydroxy-4'-methoxyisoflavone, 5,7-dihydroxy-3-4-methoxyphenyl-4h-chromen-4-one, genistein 4-methyl ether, pratensol, biochanine a, 5,7-dihydrox-4'-methoxyisoflavone, 4h-1-benzopyran-4-one, 5,7-dihydroxy-3-4-methoxyphenyl | |
CHEBI:17574 | |
COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O |
Specifications
491-80-5 | |
C16H12O5 | |
250 mg | |
biochanin a, biochanin, 4'-methylgenistein, 5,7-dihydroxy-4'-methoxyisoflavone, 5,7-dihydroxy-3-4-methoxyphenyl-4h-chromen-4-one, genistein 4-methyl ether, pratensol, biochanine a, 5,7-dihydrox-4'-methoxyisoflavone, 4h-1-benzopyran-4-one, 5,7-dihydroxy-3-4-methoxyphenyl | |
WUADCCWRTIWANL-UHFFFAOYSA-N | |
5,7-dihydroxy-3-(4-methoxyphenyl)chromen-4-one | |
5280373 | |
284.27 | |
Powder |
210°C to 213°C | |
MFCD00006839 | |
278107 | |
Soluble in water (<1mg/ml at 25°C),chloroform,methanol,DMSO (57mg/ml at 25°C),and ethanol (9mg/ml at 25°C). | |
COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O | |
284.27 | |
CHEBI:17574 | |
98% | |
5,7-Dihydroxy-4'-methoxyisoflavone |
Safety and Handling
EINECSNumber : 207-744-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only