missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5,7-Dihydroxy-4-methylcoumarin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1452806

 View more versions of this product

Catalog No. AAA1452806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5,7-Dihydroxy-4-methylcoumarin
298°C to 300°C
MFCD00016966
179133
5,7-dihydroxy-4-methylcoumarin, 5,7-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyllimetol, 4-methyl-5,7-dihydroxycoumarin, 2h-1-benzopyran-2-one, 5,7-dihydroxy-4-methyl, 5,7-dihydroxy-4-methyl-2h-1-benzopyran-2-one, coumarin, 5,7-dihydroxy-4-methyl, 5,7-dihydroxy-4-methyl-chromen-2-one, chembl12252, unii-vp62d4346m
CC1=CC(=O)OC2=CC(=CC(=C12)O)O
192.17
192.17
2107-76-8
C10H8O4
5 g
Light sensitive
QNVWGEJMXOQQPM-UHFFFAOYSA-N
5,7-dihydroxy-4-methylchromen-2-one
5354284
98%

Chemical Identifiers

2107-76-8
192.17
QNVWGEJMXOQQPM-UHFFFAOYSA-N
5354284
CC1=CC(=O)OC2=CC(=CC(=C12)O)O
C10H8O4
MFCD00016966
5,7-dihydroxy-4-methylcoumarin, 5,7-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyllimetol, 4-methyl-5,7-dihydroxycoumarin, 2h-1-benzopyran-2-one, 5,7-dihydroxy-4-methyl, 5,7-dihydroxy-4-methyl-2h-1-benzopyran-2-one, coumarin, 5,7-dihydroxy-4-methyl, 5,7-dihydroxy-4-methyl-chromen-2-one, chembl12252, unii-vp62d4346m
5,7-dihydroxy-4-methylchromen-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-289-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only