missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Acetamido-2-aminobenzoic acid, 97%, Thermo Scientific Chemicals

$77.78 - $231.92

Chemical Identifiers

CAS 50670-83-2
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD00060120
InChI Key GSOHXJQXAKNJES-UHFFFAOYSA-N
Synonym 5-acetamidoanthranilic acid, 2-amino-5-acetamidobenzoic acid, benzoic acid, 5-acetylamino-2-amino, 5-acetylamino-2-aminobenzoic acid, 5-acetamino-2-aminobenzoic acid, 5-acetamidoanthranilicacid, acmc-209kp2, 5-acetylaminoanthanilic acid, 5-acetamido-2-aminobenzoicacid, 2-amino-5-acetaminobenzoic acid
PubChem CID 170890
IUPAC Name 5-acetamido-2-aminobenzoic acid
SMILES CC(=O)NC1=CC(=C(C=C1)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1125606
View Documents
Thermo Scientific Chemicals
L1125606
5 g
Each for $77.78
 
AAL1125614
View Documents
Thermo Scientific Chemicals
L1125614
25 g
Each for $231.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50670-83-2
194.19
GSOHXJQXAKNJES-UHFFFAOYSA-N
170890
CC(=O)NC1=CC(=C(C=C1)N)C(=O)O
C9H10N2O3
MFCD00060120
5-acetamidoanthranilic acid, 2-amino-5-acetamidobenzoic acid, benzoic acid, 5-acetylamino-2-amino, 5-acetylamino-2-aminobenzoic acid, 5-acetamino-2-aminobenzoic acid, 5-acetamidoanthranilicacid, acmc-209kp2, 5-acetylaminoanthanilic acid, 5-acetamido-2-aminobenzoicacid, 2-amino-5-acetaminobenzoic acid
5-acetamido-2-aminobenzoic acid

Specifications

50670-83-2
C9H10N2O3
5 g
5-acetamidoanthranilic acid, 2-amino-5-acetamidobenzoic acid, benzoic acid, 5-acetylamino-2-amino, 5-acetylamino-2-aminobenzoic acid, 5-acetamino-2-aminobenzoic acid, 5-acetamidoanthranilicacid, acmc-209kp2, 5-acetylaminoanthanilic acid, 5-acetamido-2-aminobenzoicacid, 2-amino-5-acetaminobenzoic acid
CC(=O)NC1=CC(=C(C=C1)N)C(=O)O
194.19
194.19
5-Acetamido-2-aminobenzoic acid
∽230°C (decomposition)
MFCD00060120
2726882
GSOHXJQXAKNJES-UHFFFAOYSA-N
5-acetamido-2-aminobenzoic acid
170890
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 256-702-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only