missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide 97.0+%, TCI America™

Supplier:  TCI America A216925G

Catalog No. A216925G


Explore available promotions

Description

Description

Specifications

Specifications

5-Acetamido-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
275°C
MFCD08063354
5-Acetamido-N,N′C-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
CC(=O)NC1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
747.06
747.06
Crystalline Powder
31127-80-7
C16H20I3N3O7
25 g
BHCBLTRDEYPMFZ-UHFFFAOYNA-N
N1,N3-bis(2,3-dihydroxypropyl)-5-acetamido-2,4,6-triiodobenzene-1,3-dicarboxamide
11115326
≥97.0% (N)

Chemical Identifiers

31127-80-7
747.06
BHCBLTRDEYPMFZ-UHFFFAOYNA-N
11115326
CC(=O)NC1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
C16H20I3N3O7
MFCD08063354
5-Acetamido-N,N′C-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
N1,N3-bis(2,3-dihydroxypropyl)-5-acetamido-2,4,6-triiodobenzene-1,3-dicarboxamide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions